Cleanroom Labels, Core2Write
Core2Write label, sterile, irradiated, 50,8×25,4 mm, 36 labels/sheet/500 sheets/case
Catalog # 76628-007
Supplier: Veltek Associates
Supplier #: C2WR-2X1-SHEET-02
Jump to:
Product Details & Documents
Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
Specifications
- Description:Core2Write label, sterile, irradiated
- Dimensions:50,8×25,4 mm
- Packaging:36 labels/sheet/500 sheets/case