Methyltransferase Assay Buffer I
Buffer for enzyme assay, 5 ml
Catalog # 77559-338
Supplier: Sino Biological
Supplier #: M01-09-05
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Stock solution of the buffer used for assaying protein methyltransferases.
Storage: For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Documents
TDS Methyltransferase Assay Buffer I
Datasheet
Specifications
Specifications
- Description:Buffer for enzyme assay
- Size:5 ml
- Cat. No.:77559-338
- Formulation:Frozen solution of buffer containing: 250 mM Tris-HCl, pH 9.0 and 50 ng/µl BSA.
- Storage temperature:–20 °C



