HPLC Supply and Waste Starter Bundle PLUS, Type 20
HPLC Supply and waste starter bundle PLUS with 1×LISA funnel extension, V3.0, Thread size: GL 45, S60/61, HDPE, 20 L
Catalog # 77553-652
Supplier: SCAT AMERICAS, INC.
Supplier #: 75102
HPLC Supply and Waste Starter Bundle PLUS, Type 20
Catalog # 77553-652
Supplier: SCAT AMERICAS, INC.
Jump to:
Product Details & Documents
HPLC Supply and waste starter bundle with labelling field, V2.0, suitable for all HPLC systems.
Certifications: UN approved.
Delivery information: Supplied with 1×Safety cap III starter kit, V2.0, GL 45, 1×LISA safety waste cap, V2.0, S60/61, 1×exhaust filter M, V3.0, with splash guard and change indicator, 1×20 L canister, S60/61, PE-HD.
Specifications
- Capacity:20 L
- Description:HPLC Supply and waste starter bundle PLUS with 1×LISA funnel extension, V3.0, Thread size: GL 45, S60/61
- Material:HDPE