PERCP F806 STREPTAVIDIN 100UG
Catalog # CA77548-360
Supplier: Biolegend
Frequently Bought Together
Phosphate, Low Range
Supplier: LaMotte
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
METHYLPARABEN 500G
Supplier: VWR
Ammonium sulfate ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
VWR® Blood Bank Saline Solutions
Supplier: VWR International
Petroleum spirit, 35…60 °C, GR ACS, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
BUFFER SOLN TOT CHLORINE 473ML
Supplier: Hach
Anti-DCX Rabbit Polyclonal Antibody
Supplier: Prosci
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics
Toluene ACS
Supplier: INNOVA CHEMICALS CA
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Glycerine (natural) ≥99.0% USP
Supplier: Spectrum Chemical Mfg. Corp.