$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
ANTIBODY SPKYG 570 MU IGM Κ ITCL 25UG
Catalog # 77548-314
Supplier:
Biolegend
New Product
Expand image
ANTIBODY SPKYG 570 MU IGM Κ ITCL 25UG
Catalog # 77548-314
Supplier:
Biolegend
Supplier Number:
401637
New Product
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International