0
0,00 $
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FLUOROMETRIC ASSAY BETA NAG ACTIVITY
Catalog # MSPP-TBS2105F1
Supplier:
TRIBIOSCIENCE, INC. MS
Expand image
FLUOROMETRIC ASSAY BETA NAG ACTIVITY
Catalog # MSPP-TBS2105F1
Supplier:
TRIBIOSCIENCE, INC. MS
Supplier Number:
TBS2105F-100
Related Information
Documentation
Datasheet
TDS Beta- N-Acetylglucosaminidase (Beta - NAG) Activity Fluorometric Assay (Store at -20℃).pdf
Documentation
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma