Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ASSAY CD38 CYCLASE ACTIVITY 100 TESTS
Catalog # MSPP-TBS210010
Supplier:
TRIBIOSCIENCE, INC. MS
Expand image
ASSAY CD38 CYCLASE ACTIVITY 100 TESTS
Catalog # MSPP-TBS210010
Supplier:
TRIBIOSCIENCE, INC. MS
Supplier Number:
TBS2100-100
Related Information
Documentation
Datasheet
TDS CD38 Cyclase Activity Assay Kit.pdf
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH