$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
ASSAY CYCLIC AMP COLORIMETRIC 100 TESTS
MSPP-TBS207310
:
TRIBIOSCIENCE, INC. MS
ASSAY CYCLIC AMP COLORIMETRIC 100 TESTS
MSPP-TBS207310
:
TRIBIOSCIENCE, INC. MS
:
TBS2073-100
TDS Cyclic AMP (cAMP) Colorimetric Assay.pdf
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH