Orders LinkContactUsLinkComponent
Polyethylene Dropping Bottles
Polyethylene Dropping Bottles
29 ml (1 oz.), LDPE
Catalog # 470356-534
Supplier #:  605065-0010
Polyethylene Dropping Bottles
Catalog # 470356-534
Supplier:  
Polyethylene Dropping Bottles
Polyethylene Dropping Bottles
29 ml (1 oz.), LDPE
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BOTTLE DROPPING LDPE 1OZ Catalog #470356-535
Product Details & Documents
Economy LDPE dropper bottles.
  • Translucent LDPE dropper bottles
  • Sturdy white dropping cap
  • Flip up spout
Dropper bottles with hinged dispenser caps for dispensing indicator solutions, dyes, and more.
Specifications
  • Capacity:
    29 ml (1 oz.)
  • Material:
    LDPE