INJECTOR PORT GRAPHPACK 530UM
Catalog # AG5021-7168
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
INJECTOR PORT GRAPHPACK 530UM
Catalog # AG5021-7168
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier Number: 5021-7168
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
SEAL ALUM SLVR 20MM CS1000
Supplier: DWK Life Sciences (KIMBLE)
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
VWR® Crimp Caps for 11 mm Crimp Top Vials
Supplier: VWR International
AQ™ Advanced Quality Borosilicate Glass and Plastic Inserts
Supplier: MicroSolv Technology Corp
VWR® Inserts for Wide Opening Vials
Supplier: VWR International
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Supplier: VWR International
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
High Recovery Vials and Inserts, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
SureSTART™ MSCERT+ Screw Vial and Cap Kits, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
Closures for Storage Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
SureSTART™ Snap Caps, 11 mm, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
SureSTART™ Caps for 20 ml Glass Crimp Top Headspace Vials, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
Long Life Septa for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
DELIVERY CAP SOLID GL-45
Supplier: MicroSolv Technology Corp
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Supplier: Thermo Scientific