Orders LinkContactUsLinkComponent
QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
QuEChERS dSPE kit, TFDA method, includes animal origin, 15 ml
Catalog # AG5982-6665
Supplier #:  5982-6665
QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
Catalog # AG5982-6665
CAS Number:  
QuEChERS Dispersive Solid Phase Extraction (SPE) Kits
QuEChERS dSPE kit, TFDA method, includes animal origin, 15 ml
CAS Number:  
Price
$467.00
Log in for availability
Each(1/Itm/Bx)
Product Details & Documents
QuEChERS dispersive solid phase extraction (SPE) kits supplied in 15 ml centrifuge tubes accommodate the aliquot volumes specified by current AOAC and EN methodologies.
  • Non hazardous
The sorbent extracts the sample matrix, while MgSO₄ helps remove excess water and improve analyte partitioning.
Specifications
Specifications
  • Capacity:
    15 ml
  • Description:
    QuEChERS dSPE kit, TFDA method, includes animal origin