Inoculating Needle/Loops
Inoculating loop, 10 μl, Yellow, 400/pack, 4000/case
Catalog # 77508-610
Supplier: NEST SCIENTIFIC
Supplier #: 718201
Jump to:
Product Details & Documents
Inoculating needle/loops made up of PP material, hydrophilic.
- Applied in microbiology experiments
- Different colors for identification
- Sterilized by ethylene oxide
- Easy-tear sterile individual packaging
Flexible handles for easier collection and inoculation.
Specifications
- Color:Yellow
- Description:Inoculating loop, 10 μl
- Packaging:400/pack, 4000/case