Order Entry
Canada
Orders LinkContactUsLinkComponent
Cryo Boxes, PC

Cryo Boxes, PC

Blue, 1.9 ml vials, 8×6 array, PC
Catalog # 77508-380
Supplier:   NEST SCIENTIFIC
Supplier #:  616931
Cryo Boxes, PC
Catalog # 77508-380
Supplier:  NEST SCIENTIFIC
Cryo Boxes, PC

Blue, 1.9 ml vials, 8×6 array, PC

Price
$118.78
Log in for availability
Case of 10 Each
Product Details & Documents

Cryo boxes are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.

  • Temperature range: −196 to +70 ℃
  • Individual package
  • Clear polycarbonate cover for easy identification
  • Alphanumeric grid for easy sample identification
  • Each box includes drain holes and air vents to minimize condensation
Specifications
  • Color:
    Blue
  • Material:
    PC
  • For vials:
    1.9 ml vials, 8×6 array