Orders LinkContactUsLinkComponent
Cryo Boxes, PC
Blue, 3.0 ml vials, 9×9 array, PC
  77508-376
 :  616421
Cryo Boxes, PC
  77508-376
Cryo Boxes, PC
Blue, 3.0 ml vials, 9×9 array, PC
$531.76
Case of 32 Each
Cryo boxes are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
  • Temperature range: −196 to +70 ℃
  • Individual package
  • Clear polycarbonate cover for easy identification
  • Alphanumeric grid for easy sample identification
  • Each box includes drain holes and air vents to minimize condensation
  • Color:
    Blue
  • Material:
    PC
  • For vials:
    3.0 ml vials, 9×9 array