Order Entry
Canada
Orders LinkContactUsLinkComponent
Cryo Boxes, PC

Cryo Boxes, PC

Blue, 0.5 ml vials, internal thread cryogenic vial only, 10×10 array, PC
Catalog # 77508-368
Supplier:   NEST SCIENTIFIC
Supplier #:  616204
Cryo Boxes, PC
Catalog # 77508-368
Supplier:  NEST SCIENTIFIC
Cryo Boxes, PC

Blue, 0.5 ml vials, internal thread cryogenic vial only, 10×10 array, PC

Price
$709.02
Log in for availability
Case of 56 Each
Product Details & Documents

Cryo boxes are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.

  • Temperature range: −196 to +70 ℃
  • Individual package
  • Clear polycarbonate cover for easy identification
  • Alphanumeric grid for easy sample identification
  • Each box includes drain holes and air vents to minimize condensation
Specifications
  • Color:
    Blue
  • Material:
    PC
  • For vials:
    0.5 ml vials, internal thread cryogenic vial only, 10×10 array
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.