Orders LinkContactUsLinkComponent
Colored Cap Inserts
Colored Cap Inserts
Coloured cap insert, Blue
Catalog # 77508-332
Supplier #:  611004
Colored Cap Inserts
Catalog # 77508-332
Supplier:  
Colored Cap Inserts
Coloured cap insert, Blue
Discontinued
Product Details & Documents
Six colored cap inserts for easy identification. Compatible with regular cryogenic vials that with external thread caps only.
Packaging: 100/pack; 1000/case
Specifications
  • Color:
    Blue
  • Description:
    Coloured cap insert