Erlenmeyer Flasks, Polycarbonate
Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 12 per case, 250 ml, Sterile, Vent cap
Catalog # 77508-830
Supplier: NEST SCIENTIFIC
: 782211
Jump to:
Product Details & Documents
PC Erlenmeyer flask with flat or baffled bottom are equipped with seal caps or vent filter caps. The whole collection of Erlenmeyer flasks are sterile and enzyme-free, and are intended for the culture of suspended cells and bacteria.
- PC bottle, which meets the USP Class 6 standard, with high transparency, has strong impact resistance, oxidation resistance and can withstand a high temperature up to 121 ℃
- There is a scale made by injection molding on the flask body to facilitate observing the filled liquid volume
- Non-pyrogenic, DNase/Rnase free
- Electron beam sterilization, SAL= 10⁻⁶
- Individually packaged
In the culture process of mammalian cells and insect cells, the cells show strong viability and high protein expression in Erlenmeyer flasks, baffled. The groove at the bottom forms a vortex that accelerates the reagent flow to improve the oxygen exchange rate between the surface and the outside for higher dissolved oxygen content.
Baffle-designed erlenmeyer flasks promote optimal and consistent solution agitation when used with flask shakers.
: ISO 9001 and 13485 certified.
Specifications
- Cap type:Vent cap
- Sterility:Sterile
- Capacity:250 ml
- Description:Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 12 per case



