Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Sample Vials, with HDPE Caps

Sample vial, 5 ml, Yes, Pink capture cap, Sterile, 50/pack; 1000/case
Catalog # 77508-412
Supplier:   NEST SCIENTIFIC
 :  619201
Sample Vials, with HDPE Caps
Catalog # 77508-412
Supplier:  NEST SCIENTIFIC
Sample Vials, with HDPE Caps

Sample vial, 5 ml, Yes, Pink capture cap, Sterile, 50/pack; 1000/case

Price
$692.45
Log in for availability
Case of 1,000 Each
Product Details & Documents

Polypropylene vials, with attached or separated HDPE Caps.

  • Self-standing: the vial placed on the lab workbench steadily even without a base
  • Sterilized by E-beam, Sterility Assurance Level: SAL 10⁻⁶
  • Safe, fast, environment-friendly, no chemical residues
  • DNAse-, RNAse-, pyrogen-,endotoxin-free
  • Disposable

Excellent sealing performance, no leakage of negative pressure 95 Kpa, ensuring safe and effective air transportation.

Temperature range: Approx.–196 to 121 °C

 : Meets USP VI standard.

Specifications
  • Cap included:
    Yes
  • Cap type:
    Pink capture cap
  • Sterility:
    Sterile
  • Capacity:
    5 ml
  • Description:
    Sample vial
  • Packaging:
    50/pack; 1000/case