Process Cooling Box
Process cooling box for 12 tubes, square, 117×117×100 mm, 160 g
Catalog # 77507-406
Supplier: NEST SCIENTIFIC
Supplier #: 200102
Jump to:
Product Details & Documents
This product can used in variety of cell types, including stem cells, primary cells, cell lines and yeast. The cooling box does not require any liquid or additive, all you need is placed the cool box into the ultra-low temperature refrigerator, then it can ensure that the box's has the cooling rate of −1 ℃ per minute.
- It is a very good device for all types cell preservation
- The product uses a symmetrical design with a combination on of foam and precious alloys
- No additive and no emissions characteristics
- It is an environmental friendly products
- It does not need to replace any parts, and can be permanently used
This product is easy to use, just insert the frozen tube, placed the unit in −80 ℃ freeze environment. It can greatly improve the handling efficiency and has a good survival rate of cell preservation. The sample is safer and worry free.
It holds upto 12 tubes - 1/2 ml cryogenic vials or 1.5/2 ml centrifuge tubes.
Specifications
- Description:Process cooling box for 12 tubes, square
- Dimensions:117×117×100 mm
- Weight:160 g



