Order Entry
Canada
Orders LinkContactUsLinkComponent
Process Cooling Box

Process Cooling Box

Process cooling box for 12 tubes, Hexagonal, Side length: 108×100 mm, 120 g
Catalog # 77507-404
Supplier:   NEST SCIENTIFIC
Supplier #:  200101
Process Cooling Box
Catalog # 77507-404
Supplier:  NEST SCIENTIFIC
Process Cooling Box

Process cooling box for 12 tubes, Hexagonal, Side length: 108×100 mm, 120 g

Price
$528.09
Log in for availability
Case of 1 Each
Product Details & Documents

This product can used in variety of cell types, including stem cells, primary cells, cell lines and yeast. The cooling box does not require any liquid or additive, all you need is placed the cool box into the ultra-low temperature refrigerator, then it can ensure that the box's has the cooling rate of −1 ℃ per minute.

  • It is a very good device for all types cell preservation
  • The product uses a symmetrical design with a combination on of foam and precious alloys
  • No additive and no emissions characteristics
  • It is an environmental friendly products
  • It does not need to replace any parts, and can be permanently used

This product is easy to use, just insert the frozen tube, placed the unit in −80 ℃ freeze environment. It can greatly improve the handling efficiency and has a good survival rate of cell preservation. The sample is safer and worry free.

It holds upto 12 tubes - 1/2 ml cryogenic vials or 1.5/2 ml centrifuge tubes.

Specifications
  • Description:
    Process cooling box for 12 tubes, Hexagonal
  • Dimensions:
    Side length: 108×100 mm
  • Weight:
    120 g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.