Orders LinkContactUsLinkComponent
Serological Pipettes
Serological Pipettes
1/pack, 500/box, 3000/case
Catalog # 77507-602
Supplier:   NEST SCIENTIFIC
Supplier #:  324001
Serological Pipettes
Catalog # 77507-602
Supplier:  NEST SCIENTIFIC
Serological Pipettes
1/pack, 500/box, 3000/case
Price
885,48 $
Log in for availability
Case of 3,000 Each
Product Details & Documents
Serological pipettes can be used for liquid transfer in biological experiments such as cell culture and bacterial culture.
  • Made of high clarity polystyrene
  • Sterilized by E-beam
  • Non-pyrogenic, non-cytotoxic and non-hemolytic
  • DNase/RNase free
  • Polyolefin fiber filter to reduce contamination
  • Edged color-coded band for identification
  • Negative graduations for extra volume
Specifications
  • Packed:
    1/pack, 500/box, 3000/case