Order Entry
Canada
Orders LinkContactUsLinkComponent
Pasteur Pipettes

Pasteur Pipettes

Pasteur pipette, extra long, PP, 3 ml, 182 mm, Sterile, 2000/case
Catalog # 77507-592
Supplier:   NEST SCIENTIFIC
Supplier #:  318314
Pasteur Pipettes
Catalog # 77507-592
Supplier:  NEST SCIENTIFIC
Pasteur Pipettes

Pasteur pipette, extra long, PP, 3 ml, 182 mm, Sterile, 2000/case

Price
$413.86
Log in for availability
Case of 2,000 Each
Product Details & Documents

Pipette can be twisted for use with micro vessels and special-shaped containers.

  • Good chemical stability
  • Transparent with clear graduations for easy observation
  • High precision and good repeatability
  • Individually wrapped, sterilized by EO
  • DNase/RNase, endotoxin and pyrogenic free
Specifications
  • Sterility:
    Sterile
  • Volume:
    3 ml
  • Description:
    Pasteur pipette, extra long
  • Length:
    182 mm
  • Material:
    PP
  • Packed:
    2000/case