Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Pasteur Pipettes

Pasteur pipette, PP, 3 ml, 162 mm, Non sterile, 5000/case
  77507-590
 :  318231
Pasteur Pipettes
  77507-590
Pasteur Pipettes

Pasteur pipette, PP, 3 ml, 162 mm, Non sterile, 5000/case

 
$399.63
   
Case of 5,000 Each
 

Pipette can be twisted for use with micro vessels and special-shaped containers.

  • Good chemical stability
  • Transparent with clear graduations for easy observation
  • High precision and good repeatability
  • Individually wrapped, sterilized by EO
  • DNase/RNase, endotoxin and pyrogenic free
 
  • Sterility:
    Non sterile
  • Volume:
    3 ml
  • Description:
    Pasteur pipette
  • Length:
    162 mm
  • Material:
    PP
  • Packed:
    5000/case