Orders LinkContactUsLinkComponent
Robotic Pipette Tips
Robotic Pipette Tips
300 µl, +, 96 pieces/rack 24 racks/pack
Catalog # 77507-870
Supplier:   NEST SCIENTIFIC
Supplier #:  345613
Robotic Pipette Tips
Catalog # 77507-870
Supplier:  NEST SCIENTIFIC
Robotic Pipette Tips
300 µl, +, 96 pieces/rack 24 racks/pack
Price
$444.01
Log in for availability
Case of 2,304 Each
Product Details & Documents
NEST robotic conductive tip can be applied in R and D of genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals and other high flux liquid transfer.
  • Made of polymer material polypropylene (PP)
  • Offers high efficiency and repeatability
  • With barcode
Surface owns high hydrophobicity as treated with a special technique making for certain purposes.
Specifications
  • Volume:
    300 µl
  • Packed:
    96 pieces/rack 24 racks/pack
  • Sterile:
    +