Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Phenol red solution pH-indicator

Catalog # 104085-816
Marketsource
 :  21126
Phenol red solution pH-indicator
Catalog # 104085-816
Supplier:  
Marketsource
CAS Number:  143-74-8
Phenol red solution pH-indicator
Phenol red solution pH-indicator
CAS Number:  143-74-8
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PHENOL RED SOLN 0.02%(AQ) 100ML Catalog #470301-964

 

Product Details & Documents

pH Indicator Solution

  • Color range: pH 6.5-8.5
Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
  • Storage Temperature:Ambient