Electro-Gel™ Electrolyte Replenishers
1 oz., 60 cups/case
Catalog # CA76406-464
Supplier: Bio-Serv
Supplier #: EGS-1
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Gelled, oral electrolyte hydration supplement. Provides balanced electrolyte and glucose levels to provide energy, nutrients and hydration to accelerate recovery.
- Economical and easy to use requiring no refrigeration or special storage
- Highly palatable with a sweet, salty orange or strawberry flavor
- Comparable to Pedialyte and PRANG in gel form
- Suitable for a variety of research species
- Thermoreversible
- Gamma Irradiated
Shelf life of 12 months, cool dry storage.
Ingredients: Purified water, dextrose, gelatin, L-ascorbic acid phosphate, sodium chloride, sodium bicarbonate, citric acid, potassium chloride, flavoring, natural color, potassium sorbate, acesulfame potassium.
Specifications
- Size:1 oz.
- Packaging:60 cups/case