ReadiUse™ Annexin Binding Buffer, Optimized for Imaging and Flow Cytometry
ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 100 ml
Catalog # 77461-920
Supplier: AAT Bioquest
Supplier #: 22871
ReadiUse™ Annexin Binding Buffer, Optimized for Imaging and Flow Cytometry
Catalog # 77461-920
Supplier: AAT Bioquest
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The ReadiUse™ Annexin Binding Buffer *Optimized for Imaging and Flow Cytometry* is an isotonic buffer designed to promote the labeling of apoptotic cells with annexin V conjugates.
- Provided as a 10X concentrate
- Saves time
- pH of 7.4
Prior to annexin V staining, we ecommend diluting the 10X concentrate to a 1X working solution using distilled water.
Caution: For research use only.
Specifications
Specifications
- Description:ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 100 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



