Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RNA POLYMERASE T7 TURBO GMP GRADE 1ML
Catalog # MSPP-GMP412PB1
Supplier:
VAZYME INTERNATIONAL LLC MS
Expand image
RNA POLYMERASE T7 TURBO GMP GRADE 1ML
Catalog # MSPP-GMP412PB1
Supplier:
VAZYME INTERNATIONAL LLC MS
Supplier Number:
GMP4120PB-01
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International