BIOXP® Select mRNA
Kit for BioXp 3250, no modifications, 8
Catalog # 77461-280
Supplier: TELESIS BIO
Supplier #: BX-MRNA-RSU-CC-08
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The BioXp Select mRNA synthesis kit enables the synthesis of mRNA from an input DNA template.
- mRNA Length: 0.4 to 10 kb
- Yield: >50 µg/well
- Compatible with BioXp 3250
- Cap: CleanCap® AG (3'OMe)
- Modification: Pseudouridine
Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.
Documents
Specifications
Specifications
- Description:Kit for BioXp 3250, no modifications
- No. of reactions:8
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



