Order Entry
Canada
Orders LinkContactUsLinkComponent
BIOXP® Select mRNA

BIOXP® Select mRNA

Kit for BioXp 3250, psuedouridine modifications, 8
Catalog # 77461-274
Supplier:   TELESIS BIO
Supplier #:  BX-MRNA-RSM1-CC-08
BIOXP® Select mRNA
Catalog # 77461-274
Supplier:  TELESIS BIO
CAS Number:  
BIOXP® Select mRNA
BIOXP® Select mRNA

Kit for BioXp 3250, psuedouridine modifications, 8

CAS Number:  
Price
$2,378.38
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The BioXp Select mRNA synthesis kit enables the synthesis of mRNA from an input DNA template.

  • mRNA Length: 0.4 to 10 kb
  • Yield: >50 µg/well
  • Compatible with BioXp 3250
  • Cap: CleanCap® AG (3'OMe)
  • Modification: Pseudouridine

Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.

Specifications

Specifications

  • Description:
    Kit for BioXp 3250, psuedouridine modifications
  • No. of reactions:
    8

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.