Order Entry
Canada
Orders LinkContactUsLinkComponent
NEXTFLEX™ UDI Barcodes

NEXTFLEX™ UDI Barcodes

NEXTFLEX™ UDI barcodes (1 - 96), 192
Catalog # CA77448-162
Supplier #:  NOVA-514150
NEXTFLEX™ UDI Barcodes
Catalog # CA77448-162
CAS Number:  
NEXTFLEX™ UDI Barcodes
NEXTFLEX™ UDI Barcodes

NEXTFLEX™ UDI barcodes (1 - 96), 192

CAS Number:  
Price
$3,035.98
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The NEXTFLEX® Unique dual index barcodes are barcoded adapters that can be used with single or paired-end read sequencing library prep kits  designed for sequencing on Illumina® platforms workflows.

  • 384 UDI adapters for mitigating sample misassignment due to index hopping
  • Unique dual index adapters for multiplexing up to 384 libraries for Illumina® sequencing platforms
  • Mitigate index hopping or spread of signal that can occur during sequencing
  • Each lot of the NEXTFLEX® Unique dual index barcodes is tested for index purity by sequencing
  • Compatible with both paired-end and single-read sequencing
  • Compatible with the NEXTFLEX® Universal blockers

The NEXTFLEX® Unique dual index barcodes utilize two unique 8 nt indexes on each UDI adapter. The 8 nt indices are designed with a Hamming Distance of at least three throughout the entire series, permitting error correction. Patterned flow cells, such as those found on the Illumina® NovaSeq® and HiSeq® 3000/4000 platforms have been recognized to suffer from increased rates of sample mis-assignment during sequencing.

The use of adapters with unique dual indices during sequencing prevents such mis-assigned reads from appearing in final data sets, allowing for the highest assurance of data integrity. The NEXTFLEX® Unique dual index barcodes are barcoded adapters, for sequencing on Illumina® platforms, that provide data security in sequencing applications. Multiplexing with NEXTFLEX® Unique dual index barcodes increase scale while reducing costs by allowing the user to pool up to 384 libraries in a single flow cell lane.

Certifications: Manufactured in ISO 9001 facility.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Description:
    NEXTFLEX™ UDI barcodes (1 - 96)
  • No. of reactions:
    192

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.