BIOXP® Select Plasmid AMP
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
BioXp Select plasmid amplification kits contain reagents for isothermal amplification of plasmid DNA.
- Input: Plasmids ranging from 1.8 to 37 kb
- Yield: >10 µg/well
- Output format: Branched concatemers
- Compatible with BioXp 3250
Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.
Documents
BioXp® Select Plasmid Amplification Kit - Loading Map Checklist and Kit Information
Additional Product Details
Telesis Bio BioXp® DNA Amplification Kits
Product Literature
Specifications
Specifications
- Description:Kit for BioXp 3250
- No. of reactions:16
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)