Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Explorer™ Precision Balances

EX6202N, 6200 g, 190×200 mm (7.5×8"), 0.01 g
Catalog # 77435-008
Supplier:   Ohaus
 :  30553888
Explorer™ Precision Balances
Catalog # 77435-008
Supplier:  Ohaus
Explorer™ Precision Balances
Explorer™ Precision Balances

EX6202N, 6200 g, 190×200 mm (7.5×8"), 0.01 g

Price
$9,455.94
Log in for availability
Each
Product Details & Documents

The Explorer series is the flagship precision balance range from OHAUS. Equipped with a weighing cell that's precision-machined from a solid metal block, Explorer balances are extremely accurate and durable, and are ideal for laboratory and industrial environments.

  • Four-level user management with password protection, and un-editable system log plus a high level of configurability make Explorer balances ideal for regulated applications
  • Detachable terminal with a large color touchscreen and programmable IR sensors
  • Frameless draftshield make Explorer precision balances easy and convenient to use

The OHAUS Explorer Series of precision balances combine modern features and design elements to offer unmatched functionality in a line of high-performance balances unlike any other on the market. These smart and intuitive balances simplify even the most complex laboratory measurements.

Specifications
  • Model:
    EX6202N
  • Pan/Platform Size:
    190×200 mm (7.5×8")
  • Readability:
    0.01 g
  • Weighing capacity:
    6200 g