Orders LinkContactUsLinkComponent
Accessories for Compresstome® Buffer Tray
Compresstome stainless steel buffer tray
  77435-448
 :  VFBTVMSSBOS
Accessories for Compresstome® Buffer Tray
  77435-448
Accessories for Compresstome® Buffer Tray
Accessories for Compresstome® Buffer Tray
Compresstome stainless steel buffer tray
$839.85
Each
Stainless steel Compresstome buffer tray is typically used for sterile sectioning procedures.
  • Buffer tray is autoclavable
  • Full one year warranty
Replacement gasket and screws set for the buffer tray for Compresstome tissue slicers. This set includes one set of screws and one gasket.
Compresstome buffer tray made of high-quality, durable stainless steel. Autoclavable. Full one year warranty. This set includes, one stainless steel buffer tray, one set of screws, one gasket, and one small Allen wrench.
  • Description:
    Compresstome stainless steel buffer tray
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.