Compresstome® Sectioning Tools
Product Details & Documents
Sectioning tools designed for use with compresstome rotary microtomes.
- Fine tissue forceps are supplied individually
- Tissue paintbrushes are supplied in packs of 3
Fine tissue handling forceps enable gentle manipulation of tissues and organs during dissection. They allow for the gentle removal of the tissue from the bufffer tray while slicing.
Tissue paintbrushes allow for easy handling of delicate tissue slices. Soft bristles help prevent damage to tissue sections when transferring slices.
Specifications
- Description:Fine tissue handling forceps
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)