Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GLOVE PORT HP BUTYL 10 24 ML 32 8.5 AB
Catalog # 77400-976
Supplier:
PIERCAN USA INC
Expand image
GLOVE PORT HP BUTYL 10 24 ML 32 8.5 AB
Catalog # 77400-976
Supplier:
PIERCAN USA INC
Supplier Number:
10BHP2432-8.5A
Frequently Bought Together
VWR® Lab Markers
Supplier: VWR International
VWR® General Purpose Laboratory Labeling Tape
Supplier: VWR International
Alcohol- and Water-Resistant Cryogenic Markers, Fine Tip
Supplier: GA INTERNATIONAL INC
Color Lab Tape
Supplier: GA INTERNATIONAL INC
VWR® Cryogenic Dot and Strip Labels
Supplier: VWR International
VWR® Lead-Free Autoclave Tapes
Supplier: VWR International
Cryogenic Color Dots Writable Labels
Supplier: GA INTERNATIONAL INC