0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GLYCOPROTEIN PRE-FUSION F0 RSFV52H8500UG
Catalog # 104049-624
Supplier:
ACROBIOSYSTEMS INC MS
Expand image
GLYCOPROTEIN PRE-FUSION F0 RSFV52H8500UG
Catalog # 104049-624
Supplier:
ACROBIOSYSTEMS INC MS
Supplier Number:
RSFV52H8500UG
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning