BioX Cryogenic Vials, Barcoded
Barcoded cryovial, 1D/Human readable and 2D on bottom, 5.0 ml, Purple, Sterile
77395-070
: BX88-9505
Barcoded polypropylene cryogenic vials with colored caps.
- Suitable for liquid nitrogen storage
- Pre-barcoded
- O-ring free
- Self standing
- White writing patch and graduations
Unique permanent barcodes for superior traceability. 1D and human-readable on the side are perfect for hand scanning and the 2D barcodes on the bottom make these vials automation friendly.
: Use in liquid phase LN₂ is not recommended.
- Sterility:Sterile
- Capacity:5.0 ml
- Description:Barcoded cryovial, 1D/Human readable and 2D on bottom
- Cap color:Purple