BioX Cryogenic Vials, Barcoded
Barcoded cryovial, 1D/Human readable, internal thread, 2.0 ml, Purple, Sterile
Catalog # 77395-004
Supplier: TWD Scientific
: BX88-6215
Jump to:
Product Details & Documents
Barcoded polypropylene cryogenic vials with colored caps.
- Suitable for liquid nitrogen storage
- Pre-barcoded
- O-ring free
- Self standing
- White writing patch and graduations
Unique permanent barcodes for superior traceability. 1D and human-readable on the side are perfect for hand scanning and the 2D barcodes on the bottom make these vials automation friendly.
: Use in liquid phase LN₂ is not recommended.
Specifications
- Sterility:Sterile
- Capacity:2.0 ml
- Description:Barcoded cryovial, 1D/Human readable, internal thread
- Cap color:Purple