$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
LAB COATS WH S CS50
77393-986
:
APEX ASEPTIC PRODUCTS, LLC.
LAB COATS WH S CS50
77393-986
:
APEX ASEPTIC PRODUCTS, LLC.
:
3460WHS
Frequently Bought Together
VWR® Lab Markers
: VWR International
VWR® General Purpose Laboratory Labeling Tape
: VWR International
Color Lab Tape
: GA INTERNATIONAL INC
VWR® Lead-Free Autoclave Tapes
: VWR International
Charts and Pens for 6" Dickson® Chart Recorders, Graphic Controls
: Graphic Controls
Rainbow Laboratory Tape
: NEVS INK INC SE
VWR® Cryogenic Dot and Strip Labels
: VWR International