Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SCRUB TOP SMS BLUE S CS50.
Catalog # 77393-998
Supplier:
APEX ASEPTIC PRODUCTS, LLC.
Expand image
SCRUB TOP SMS BLUE S CS50.
Catalog # 77393-998
Supplier:
APEX ASEPTIC PRODUCTS, LLC.
Supplier Number:
3520LE-CBS
Frequently Bought Together
VWR® Lab Markers
Supplier: VWR International
VWR® Labels 'Radioactive Warning'
Supplier: VWR International
Color Lab Tape
Supplier: GA INTERNATIONAL INC
VWR® General Purpose Laboratory Labeling Tape
Supplier: VWR International
VWR® Multiple Roll Tape Dispensers
Supplier: VWR International
BRAND® UV-Cuvette Disposable Spectrophotometer/Photometer Cuvettes, BrandTech
Supplier: Brandtech
Accessories for VWR® Large Shelf Label Holders
Supplier: VWR International
Cryogenic Tapes for Laboratory Use
Supplier: GA INTERNATIONAL INC