$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
LAB COAT WH XL CS100
77393-988
:
APEX ASEPTIC PRODUCTS, LLC.
LAB COAT WH XL CS100
77393-988
:
APEX ASEPTIC PRODUCTS, LLC.
:
3460WHXL
Frequently Bought Together
VWR® Lab Markers
: VWR International
PTFE Tape, Chemglass
: Chemglass
Single Roll Tape Dispenser
: NEVS INK INC SE
Color Lab Tape
: GA INTERNATIONAL INC
VWR® General Purpose Laboratory Labeling Tape
: VWR International
VWR® Cryogenic Dot and Strip Labels
: VWR International
Cryogenic Tapes for Laboratory Use
: GA INTERNATIONAL INC
VWR® Stat Labels
: VWR International
Labels 'Hazard and Handling', Sonoco
: Sonoco Thermosafe