Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
GOWN ISO KNIT CUFF BLUE REG CS50
77393-946
:
APEX ASEPTIC PRODUCTS, LLC.
GOWN ISO KNIT CUFF BLUE REG CS50
77393-946
:
APEX ASEPTIC PRODUCTS, LLC.
:
3260B
Frequently Bought Together
VWR® Lead-Free Autoclave Tapes
: VWR International
VWR® Lab Markers
: VWR International
Color Lab Tape
: GA INTERNATIONAL INC
VWR® General Purpose Laboratory Labeling Tape
: VWR International
VWR® Laboratory Notebooks
: VWR International
Cryogenic Color Dots Writable Labels
: GA INTERNATIONAL INC