Order Entry
Canada
Orders LinkContactUsLinkComponent
PureCube 1-Step Batch MIDI Plus

PureCube 1-Step Batch MIDI Plus

48 columns
Catalog # 77393-430
Supplier:   Cube Biotech
Supplier #:  63208
PureCube 1-Step Batch MIDI Plus
Catalog # 77393-430
Supplier:  Cube Biotech
CAS Number:  
PureCube 1-Step Batch MIDI Plus
PureCube 1-Step Batch MIDI Plus

48 columns

CAS Number:  
Price
$677.67
Log in for availability
Pack of 48
Minimum order quantity is 1
Product Details & Documents

Empty 1-step batch columns are ideally suited for batch purifications. They can be filled with your preferred affinity resin, allowing binding, washing, and elution in one spin column.

  • Usage: Specific binding and purification of proteins
  • Specificity: Depending on the affinity of the used agarose resin
  • Binding capacity: Also depending on the used agarose
  • Body material: Polypropylene
  • Filter pore size: 0.1 to 0.2 µm low binding PVDF
  • Max. volume: 20 ml (midi)
  • Max. g force: 1500 to 2000×g (swing-bucket rotor)
  • Min. g force: 100 to 200×g for 1 to 10 min

Specifically designed for small to mid-scale protein purifications, PureCube 1-step batch spin columns save time and pipetting steps. Containing a SelfSeal membrane technology, the column retains resin and sample in a chamber for batch incubation. By centrifugation, the membrane pores dilate, and the filtered eluate gathers in the collection chamber of the column. PureCube 1-step batch spin columns are available in two sizes: Mini for expression trials, small-scale screening, and other small-volume purification needs or Midi Plus for up to 20 ml volumes.

Required equipment:
-Lysis buffer
-Wash buffer
-Elution buffer
-Ice bath
-Refrigerated centrifuge for 50 ml tube (min 10000×g)
-50 ml centrifuge tube
-Micropipettor and micropipetting tips
-50 ml conical tubes (e.g. Falcon)
- pH meter
-End-over-end shaker
-SDS-PAGE buffers, reagents and equipment optional: Western blot reagents and equipment

Specifications

Specifications

  • Size:
    48 columns

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.