WHEATON® Serum Vials
Serum vial, 10R, clear, EBB, ISO 8362-1, EP, tray, 13.5 ml (brimful capacity), 24×45 mm, Glass, Clear, No
Supplier #: WEBB10R-T-EP
Serum vials converted from Type I borosilicate glass tubing.
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
- Crimp cap type
Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.
- Cap included:No
- Capacity:13.5 ml (brimful capacity)
- Color:Clear
- Description:Serum vial, 10R, clear, EBB, ISO 8362-1, EP, tray
- Material:Glass
- Diameter ×H:24×45 mm



