Orders LinkContactUsLinkComponent
VWR® Glove Liners
VWR® Glove Liners
Full Finger Liners, with Purple Cuff, 2X-Large
Catalog # 77392-160
Supplier:   VWR International
VWR® Glove Liners
Catalog # 77392-160
Supplier:  VWR International
VWR® Glove Liners
VWR® Glove Liners
Full Finger Liners, with Purple Cuff, 2X-Large
Price
$88.48
New Customer Promotional Price
Log in for availability
Pack of 20 Case of 300
Product Details & Documents
Ergonomically designed glove liners wick away moisture for comfortable use with disposable gloves.
  • Ambidextrous glove liners
  • Reduces perspiration and wicks away moisture
  • Low-lint, 100% 15 denier nylon composition
  • Sturdy, launderable, and reusable
  • Color-coded cuffs indicate size
  • Pre-laundered; will not shrink
Liners provide protection and comfort when used with latex, nitrile, and PVC gloves. Fully launderable for reusability and economy.
 : Glove Liners are packaged 20 pair per bag, 15 bags per case. They are packed in clean polyethylene bags that are free of silicone, dioctyl phthalate (DOP), and amides.
Specifications
  • Glove size:
    2X-Large
  • Description:
    Full Finger Liners, with Purple Cuff
  • Cat. No.:
    77392-160
  • Disposable / reusable:
    Reusable
  • Ambidextrous/Hand-specific:
    Ambidextrous
  • Cuff style:
    Knit wrist