Order Entry
Canada
Orders LinkContactUsLinkComponent
PROKLEAN BOTTLE CONTEC CAPPED 5L

PROKLEAN BOTTLE CONTEC CAPPED 5L

Catalog # 77392-230
Supplier:   Contec
Supplier #:  FBC170PK
PROKLEAN BOTTLE CONTEC CAPPED 5L
Catalog # 77392-230
Supplier:  Contec
CAS Number:  
PROKLEAN BOTTLE CONTEC CAPPED 5L
CAS Number:  
Price
$176.30
New Customer Promotional Price
Log in for availability
Each(5l) Case of 2 (5 l)
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.