WHEATON® Serum Vials
Serum vial, 3 ml, USBB, clear, EP, case, 3 ml, 16.75×37.7 mm, Glass, Clear, No
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: W0UBB3MLC-C-EP
Jump to:
Product Details & Documents
Serum vials converted from Type I borosilicate glass tubing.
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
- Crimp cap type
Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.
Documents
Specifications
- Cap included:No
- Capacity:3 ml
- Color:Clear
- Description:Serum vial, 3 ml, USBB, clear, EP, case
- Material:Glass
- Diameter ×H:16.75×37.7 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



