WHEATON® Serum Vials
Serum vial, 13.5 ml, USBB, clear, EP, pallet, 13.5 ml (brimful capacity), 24×45 mm, Glass, Clear, No
Catalog # 77392-032
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: W0EBB10R-P-EP
Jump to:
Product Details & Documents
Serum vials converted from Type I borosilicate glass tubing.
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
- Crimp cap type
Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.
Documents
Specifications
- Cap included:No
- Capacity:13.5 ml (brimful capacity)
- Color:Clear
- Description:Serum vial, 13.5 ml, USBB, clear, EP, pallet
- Material:Glass
- Diameter ×H:24×45 mm



