VWR® Disposable Cleanroom Socks
Catalog # 77322-586
Supplier: VWR International
New Product
Specifications
- Clothing Size:One size
- Garment application:Clinical,Cleanroom,Laboratory
- Color:White
- Description:VWR® Disposable cleanroom socks
- Fabric:Poly/Nylon blend
- Cat. no.:77322-586
Specifications
About this item
VWR® Disposable cleanroom socks are designed to provide all day comfort inside a cleanroom. They are made from materials that prevent any particle generation while in use.
- Single-use disposable socks
- Poly/Nylon blend
- Comfortable for extended use
- Cleanroom compatible
- Annex 1 compliant
Cleanroom socks provide comfort and durability for limited and extended wear in cleanroom environments. Suitable for use in ISO 4 to 6 (Class 10 to 1000) cleanrooms. Available in either launderable or disposable options.
Packaging: 400 pairs per case.
Frequently Bought Together